Anti-CNNM1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23655
10708-434EA 527.69 USD
10708-434
Anti-CNNM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CNNM1.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Western Blot: 1:250-1:500

Type: Primary
Antigen: CNNM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR