Anti-NUTM1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23657
10703-670EA 527.69 USD
10703-670
Anti-NUTM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C15orf55.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: NUTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR