Anti-HTR1E Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16399
10684-962EA 653.85 USD
10684-962
Anti-HTR1E Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of HTR1E.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 16-24 ?g/ml

Type: Primary
Antigen: HTR1E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Guinea Pig, Horse, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR