Anti-IFNG Mouse Monoclonal Antibody (Biotin) [clone: B27]

Supplier: Abnova
MAB5865
10735-504EA 426.15 USD
10735-504
Anti-IFNG Mouse Monoclonal Antibody (Biotin) [clone: B27]
Antibodies
Mouse monoclonal antibody raised against recombinant IFNG.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: B27
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR