Anti-IL3 Rat Monoclonal Antibody (Biotin) [clone: BVD3-1F9]

Supplier: Abnova
MAB5867
10735-254EA 527.69 USD
10735-254
Anti-IL3 Rat Monoclonal Antibody (Biotin) [clone: BVD3-1F9]
Antibodies
Rat monoclonal antibody raised against recombinant IL3.

Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: BVD3-1F9
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR