Anti-ADGRE3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16404
10683-930EA 653.85 USD
10683-930
Anti-ADGRE3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of EMR3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.5 ?g/ml

Type: Primary
Antigen: ADGRE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR