Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: JES1-39D10]

Supplier: Abnova
MAB5870
10735-270EA 527.69 USD
10735-270
Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: JES1-39D10]
Antibodies
Rat monoclonal antibody raised against recombinant IL5.

Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: JES1-39D10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR