Anti-TICAM2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13794
10686-326EA 786.15 USD
10686-326
Anti-TICAM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TICAM2.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TICAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR