Anti-IL2 Rat Monoclonal Antibody (Biotin) [clone: JES6-5H4]

Supplier: Abnova
MAB5876
10735-230EA 426.15 USD
10735-230
Anti-IL2 Rat Monoclonal Antibody (Biotin) [clone: JES6-5H4]
Antibodies
Rat monoclonal antibody raised against recombinant Il2.

Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR