Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: TRFK4]

Supplier: Abnova
MAB5878
10735-278EA 426.15 USD
10735-278
Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: TRFK4]
Antibodies
Rat monoclonal antibody raised against native Il-5.

Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: TRFK4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR