Anti-FN14 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13804
10714-968EA 1178.46 USD
10714-968
Anti-FN14 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant Fn14.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry: 2.5 ?g/ml

Type: Primary
Antigen: Fn14
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR