Anti-TP53INP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13806
10742-504EA 1178.46 USD
10742-504
Anti-TP53INP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TP53INP1.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 2 ?g/ml

Type: Primary
Antigen: TP53INP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR