Anti-IL10 Rat Monoclonal Antibody (Biotin) [clone: JES5-16E3]

Supplier: Abnova
MAB5881
10735-198EA 426.15 USD
10735-198
Anti-IL10 Rat Monoclonal Antibody (Biotin) [clone: JES5-16E3]
Antibodies
Rat monoclonal antibody raised against recombinant Il10.

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: JES5-16E3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR