Anti-TNFSF11 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13808
10712-982EA 1178.46 USD
10712-982
Anti-TNFSF11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TNFSF11.

Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: TNFSF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR