Anti-IL17A Rat Monoclonal Antibody (Biotin) [clone: TC11-8H4]

Supplier: Abnova
MAB5883
10735-352EA 527.69 USD
10735-352
Anti-IL17A Rat Monoclonal Antibody (Biotin) [clone: TC11-8H4]
Antibodies
Rat monoclonal antibody raised against recombinant Il17a.

Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: TC11-8H4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR