Anti-ZAP70 Mouse Monoclonal Antibody (Biotin) [clone: SBZAP]

Supplier: Abnova
MAB5891
10745-014EA 406.15 USD
10745-014
Anti-ZAP70 Mouse Monoclonal Antibody (Biotin) [clone: SBZAP]
Antibodies
Mouse monoclonal antibody raised against partial recombinant ZAP70.

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: SBZAP
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR