Anti-Saccharomyces cerevisiae Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13822
10628-692EA 1178.46 USD
10628-692
Anti-Saccharomyces cerevisiae Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Saccharomyces cerevisiae.

Type: Primary
Antigen: Saccharomyces cerevisiae
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR