Anti-FN1 Mouse Monoclonal Antibody [clone: Fn-3]

Supplier: Abnova
MAB7201
10714-960EA 786.15 USD
10714-960
Anti-FN1 Mouse Monoclonal Antibody [clone: Fn-3]
Antibodies
Mouse monoclonal antibody raised against recombinant FN1.

Type: Primary
Antigen: FN1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: Fn-3
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR