Anti-Streptococcus Group B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13832
10628-834EA 1440 USD
10628-834
Anti-Streptococcus Group B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Streptococcus Group B.

Type: Primary
Antigen: Streptococcus Group B
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Streptococcus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR