Anti-Pancreatic duct cell marker Mouse Monoclonal Antibody [clone: DHIC2-5H10]

Supplier: Abnova
MAB7224
10728-416EA 653.85 USD
10728-416
Anti-Pancreatic duct cell marker Mouse Monoclonal Antibody [clone: DHIC2-5H10]
Antibodies
Mouse monoclonal antibody raised against pancreatic duct cell marker.

Recommended Dilutions: Flow Cytometry: 1:50-1:100; Immunohistochemistry (Frozen sections): 1:100

Type: Primary
Antigen: Pancreatic duct cell marker
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: DHIC2-5H10
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR