Anti-Thyroid-stimulating hormone Goat Polyclonal Antibody

Supplier: Abnova
PAB13841
10629-024EA 406.15 USD
10629-024
Anti-Thyroid-stimulating hormone Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against TSH.

Recommended Dilutions: Radio Immunoassay: 1:650000

Type: Primary
Antigen: Thyroid-stimulating hormone
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR