Anti-NR0B1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2545
10682-232EA 406.15 USD
10682-232
Anti-NR0B1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of NR0B1.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 10-100

Type: Primary
Antigen: NR0B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR