Anti-CDC6 Mouse Monoclonal Antibody [clone: Cdc6 9H8/5]

Supplier: Abnova
MAB7254
10707-170EA 786.15 USD
10707-170
Anti-CDC6 Mouse Monoclonal Antibody [clone: Cdc6 9H8/5]
Antibodies
Mouse monoclonal antibody raised against full length cdc6.

Type: Primary
Antigen: CDC6
Clonality: Monoclonal
Clone: Cdc6 9H8/5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR