Anti-OLFM1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2559
10679-514EA 406.15 USD
10679-514
Anti-OLFM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Olfm1.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:60

Type: Primary
Antigen: OLFM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR