Anti-CDC37 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15584
10707-088EA 653.85 USD
10707-088
Anti-CDC37 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CDC37.

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CDC37
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR