Anti-SWI6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15585
10692-132EA 426.15 USD
10692-132
Anti-SWI6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant SWI6.

Recommended Dilutions: Western Blot: 1:2000-1:10000

Type: Primary
Antigen: SWI6
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR