Anti-PTPRC Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: EM-05]

Supplier: Abnova
MAB5119
10730-738EA 426.15 USD
10730-738
Anti-PTPRC Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: EM-05]
Antibodies
Rat monoclonal antibody raised against native Ptprc.

Recommended Dilutions: Flow Cytometry: 4 ?g/ml

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: EM-05
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR