Anti-sgo1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15588
10737-010EA 527.69 USD
10737-010
Anti-sgo1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant Sgo1.

Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: sgo1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: S. pombe
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR