Anti-CD9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: EM-04]

Supplier: Abnova
MAB5121
10706-986EA 426.15 USD
10706-986
Anti-CD9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: EM-04]
Antibodies
Rat monoclonal antibody raised against native Cd9.

Recommended Dilutions: Flow Cytometry: 6 ?g/ml

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: EM-04
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR