Anti-DNMT1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15590
10683-812EA 527.69 USD
10683-812
Anti-DNMT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant Dnmt1.

Recommended Dilutions: Western Blot: 0.2-1 ?g/ml

Type: Primary
Antigen: DNMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Frog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR