Anti-CK4, C5, C6, C8, C10, C13, and C18. Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C-11]

Supplier: Abnova
MAB5131
10709-946EA 406.15 USD
10709-946
Anti-CK4, C5, C6, C8, C10, C13, and C18. Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C-11]
Antibodies
Mouse monoclonal antibody raised against native cytokeratin.

Recommended Dilutions: Flow Cytometry: 5 ?g/ml

Type: Primary
Antigen: CK4, C5, C6, C8, C10, C13, and C18.
Clonality: Monoclonal
Clone: C-11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR