Anti-CK18 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: DC-10]

Supplier: Abnova
MAB5135
10722-888EA 406.15 USD
10722-888
Anti-CK18 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: DC-10]
Antibodies
Mouse monoclonal antibody raised against native KRT18.

Recommended Dilutions: Flow Cytometry: 5-10 ?g/ml; Immunofluorescence(5-10 ?g/ml

Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: DC-10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR