Anti-ARG1 Goat Polyclonal Antibody

Supplier: Abnova
PAB15607
10690-010EA 527.69 USD
10690-010
Anti-ARG1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of ARG1.

Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.01-0.03 ?g/ml

Type: Primary
Antigen: ARG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR