Anti-SLC39A8 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23353
10738-608EA 527.69 USD
10738-608
Anti-SLC39A8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant SLC39A8.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Western Blot: 1:250-1:500

Type: Primary
Antigen: SLC39A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR