Anti-ATG4D Goat Polyclonal Antibody

Supplier: Abnova
PAB15610
10690-392EA 527.69 USD
10690-392
Anti-ATG4D Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of ATG4D.

Recommended Dilutions: ELISA: 1:32000

Type: Primary
Antigen: ATG4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR