Anti-GTSF1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23356
10716-758EA 527.69 USD
10716-758
Anti-GTSF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant GTSF1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: GTSF1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR