Anti-CDKAL1 Goat Polyclonal Antibody

Supplier: Abnova
PAB15614
10707-342EA 527.69 USD
10707-342
Anti-CDKAL1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of CDKAL1.

Recommended Dilutions: ELISA: 1:64000

Type: Primary
Antigen: CDKAL1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR