Anti-APP Mouse Monoclonal Antibody [clone: OM16]

MAB5145
10692-770EA 1167.69 USD
10692-770
Anti-APP Mouse Monoclonal Antibody [clone: OM16]
Mouse monoclonal antibody raised against synthetic peptide of Amyloid-beta.

Recommended Dilutions: ELISA: 1 ?g/ml; Western Blot: 10 ?g/ml

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: OM16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR