Anti-ANP32A Mouse Monoclonal Antibody [clone: RJ1]

Supplier: Abnova
MAB5147
10689-224EA 725 USD
10689-224
Anti-ANP32A Mouse Monoclonal Antibody [clone: RJ1]
Antibodies
Mouse monoclonal antibody raised against recombinant ANP32A.

Recommended Dilutions: Western Blot: 1:4000; Immunocytochemistry: 1:100-1:200

Type: Primary
Antigen: ANP32A
Clonality: Monoclonal
Clone: RJ1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR