Anti-Thymine dimer photoproducts (6-4) Monoclonal Antibody [clone: KTM50]

MAB5152
10741-254EA 910.77 USD
10741-254
Anti-Thymine dimer photoproducts (6-4) Monoclonal Antibody [clone: KTM50]
Mouse monoclonal antibody raised against Thymine dimer photoproducts (6-4).

Type: Primary
Antigen: Thymine dimer photoproducts (6-4)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: KTM50
Epitope:
Host: IgG1
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR