Anti-TIMP4 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SB30c]

Supplier: Abnova
MAB5896
10628-906EA 653.85 USD
10628-906
Anti-TIMP4 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SB30c]
Antibodies
Mouse monoclonal antibody raised against recombinant TIMP4.

Type: Primary
Antigen: TIMP4
Clonality: Monoclonal
Clone: SB30c
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR