Anti-GPALPP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23373
10721-864EA 527.69 USD
10721-864
Anti-GPALPP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant KIAA1704.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: KIAA1704
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR