Anti-Fibrillin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11C1.3]

Supplier: Abnova
MAB5902
10676-268EA 786.15 USD
10676-268
Anti-Fibrillin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11C1.3]
Antibodies
Mouse monoclonal antibody raised against native Fibrillin.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: Fibrillin
Clonality: FITC (Fluorescein Isothiocyanate)
Clone: Monoclonal
Conjugation: 11C1.3
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR