Anti-PDE7B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16423
10685-734EA 653.85 USD
10685-734
Anti-PDE7B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PDE7B.

Type: Primary
Antigen: PDE7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR