Anti-PDE8A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16424
10685-736EA 653.85 USD
10685-736
Anti-PDE8A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PDE8A.

Type: Primary
Antigen: PDE8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gorilla, Horse, Human, Mouse, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR