Anti-CD3E Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 500-A2]

Supplier: Abnova
MAB5912
10734-732EA 426.15 USD
10734-732
Anti-CD3E Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 500-A2]
Antibodies
Hamster monoclonal antibody raised against Cd3e.

Recommended Dilutions: Flow Cytometry: 3 ?g/10^6 cells

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: 500-A2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype: IgG kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR