Anti-CD3E Hamster Monoclonal Antibody (Cy5®) [clone: 500-A2]

Supplier: Abnova
MAB5917
10706-428EA 406.15 USD
10706-428
Anti-CD3E Hamster Monoclonal Antibody (Cy5®) [clone: 500-A2]
Antibodies
Hamster monoclonal antibody raised against Cd3e.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: 500-A2
Conjugation: Cy5®
Epitope:
Host: Hamster
Isotype: IgG kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR