Anti-GRXCR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23701
10720-702EA 527.69 USD
10720-702
Anti-GRXCR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant GRXCR1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: GRXCR1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR