Anti-PDE6A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16433
10685-726EA 653.85 USD
10685-726
Anti-PDE6A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PDE6A.

Type: Primary
Antigen: PDE6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Hamster, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR