Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy5.5®) [clone: GK1.5]

Supplier: Abnova
MAB5925
10706-478EA 406.15 USD
10706-478
Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy5.5®) [clone: GK1.5]
Antibodies
Rat monoclonal antibody raised against Cd4.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: APC (Allophycocyanin)/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR